Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOLA1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988549
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8549 Supplier Bioss Supplier No. BS8421R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CGI-143 encodes proteins that seem to be involved in cell proliferation or cell-cycle regulation, but the molecular function is still unknown.

Specifications

Antigen BOLA1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BOLA1
Gene Accession No. Q9Y3E2
Gene Alias 1810037G04Rik; bolA family member 1; bolA homolog 1; bolA homolog 1 (E. coli); Bola1; bolA-like 1; bolA-like 1 (E. coli); BolA-like protein 1; BOS_2924; CGI-143; hBolA; LOC100357656; RGD1310586; zgc:101119
Gene Symbols BOLA1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BOLA1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51027
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.