Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BOLA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561286
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561286 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561286 Supplier Invitrogen™ Supplier No. PA561286
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKPLLQRHRL VNACLAEELP HIHAFEQKTL TPDQWARERQ K Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 88%.

This gene is located within a region of a segmental duplication on chromosome 16 and is identical to BOLA2B (bolA family member 2B). The product of this gene belongs to a family of proteins that are widely conserved and may be involved in iron maturation. Related pseudogenes are found multiple different chromosomes. Alternative splicing results in multiple transcript variants. Transcripts initiating at this locus may extend into downstream SMG1 pseudogene 6 (SMG1P6) and encode fusion proteins with a C-terminus related to SMG1 phosphatidylinositol 3-kinase-related kinase. A readthrough locus is represented with GeneID:107282092.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BOLA2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.40 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BOLA2
Gene Accession No. Q9H3K6
Gene Alias 1110025L05Rik; bolA family member 2; bolA family member 2B; bolA homolog 2; bolA homolog 2B; BOLA2; BolA-2; BOLA2A; BOLA2B; bolA-like 2 (E. coli); bolA-like protein 2; BolA-like protein 2 member A; BolA-like protein 2 member B; bolA-like protein 2B; My016; RGD1565363; similar to RIKEN cDNA 1110025L05
Gene Symbols BOLA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BOLA2. Recombinant protein control fragment (Product #RP-104406).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 552900
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.