Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BOLA3 Antibody (PCRP-BOLA3-1A5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320345100UG
Description
BOLA3 Monoclonal antibody specifically detects BOLA3 in Human samples. It is validated for Flow Cytometry, ImmunoprecipitationSpecifications
BOLA3 | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 μg/million cells, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate | |
bolA homolog 3 (E. coli), bolA-like 3, bolA-like protein 3 | |
Recombinant fragment (around aa1-103) of human BOLA3 protein (exact sequence is proprietary) | |
100 μg | |
Endocrinology, Signal Transduction | |
388962 | |
Store at 4&drg;C. Do not freeze. | |
IgG1 |
Flow Cytometry, Immunoprecipitation | |
PCRP-BOLA3-1A5 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction