Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boll Antibody (OTI2D11), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272180F
Description
Boll Monoclonal specifically detects Boll in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
Boll | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
BOLL | |
Full length recombinant protein of human Boll (NP_149019) produced in E.coli. | |
0.1 mL | |
66037.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI2D11 | |
Immunohistochemistry | |
bol, boule-like (Drosophila), BOULEbol (Drosophila boule homolog)-like, protein boule-like | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction