Learn More
Invitrogen™ BOLL Monoclonal Antibody (2G4)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA522546
Catalog No. PIMA522546
Only null left
Add to Cart
Description
Description
Peptide Sequence: FETQEDAQKI LQEAEKLNYK DKKLNIGPAI RKQQVGIPRS SIMPAAGTMY LTTSTGYPYT YHNGVAYFHT PEVTSVPPPW PSRSVCSSPV MVAQPIY.
This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BOLL
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q8N9W6
BOLL
BOLL (NP_149019, 80 a.a. ~ 176 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Immunoprecipitation, Western Blot
2G4
Unconjugated
BOLL
4930554P13Rik; 4930597B14Rik; bol, boule-like; bol, boule-like (Drosophila); Boll; BOULE; boule homolog, RNA binding protein; boule-like RNA binding protein; boule-like RNA-binding protein; protein boule-like; RGD1559527
Mouse
Affinity chromatography
RUO
66037
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.