Learn More
BOLL Mouse anti-Human, Clone: OTI5A2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804236

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
BOLL | |
Monoclonal | |
Unconjugated | |
BOLL | |
BOULE | |
Mouse | |
Affinity Chromatography | |
RUO | |
66037 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5A2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8N9W6 | |
BOLL | |
Full length human recombit protein of human BOLL produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.