Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BOLL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583929 Supplier Invitrogen™ Supplier No. PA583929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTFETQEDAQ KILQEAEKLN YKDKKLNIGP AIRKQQVGIP RSSIMPAAGT MYLTTSTGYP YTYHNGVAYF HTPEVTSVPP PWPSRSVCSS PVMVAQPIYQ Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 94%.

This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BOLL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BOLL
Gene Accession No. Q8N9W6
Gene Alias 4930554P13Rik; 4930597B14Rik; bol, boule-like; bol, boule-like (Drosophila); Boll; BOULE; boule homolog, RNA binding protein; boule-like RNA binding protein; boule-like RNA-binding protein; protein boule-like; RGD1559527
Gene Symbols BOLL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BOLL. Recombinant protein control fragment (Product #RP-92305).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66037
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.