Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bolton-Hunter Reagent, Thermo Scientific Chemicals

Catalog No. p-7082408
Click to view available options
Quantity:
1 g

For iodination of proteins that do not contain tyrosine residues

Bolton-Hunter Reagent is used for iodination of proteins that do not contain tyrosine residues, Bolton-Hunter reagent precursor (3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester) is a hydroxysuccinimide ester used as an intermediate for high radioactive radioiodination of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34071-95-9
Molecular Formula C13H13NO5, C13H13NO5
Synonym N-Succinimidyl-3-(4-hydroxyphenyl)propionate; Rudinger reagent

Specifications

For Use With (Application) For iodination of proteins that do not contain tyrosine residues
Molecular Weight (g/mol) 263.25g/mol, 263.25
Product Type Labeling Reagent
Quantity 1 g
Melting Point 132° to 133°C
Content And Storage −30° to −10°C
Beilstein 1545011
InChI Key KYRUKRFVOACELK-UHFFFAOYSA-N
SMILES OC1=CC=C(CCC(=O)ON2C(=O)CCC2=O)C=C1
IUPAC Name 2,5-dioxopyrrolidin-1-yl 3-(4-hydroxyphenyl)propanoate
Color White to Off-white
Show More Show Less
EINECSNumber 251-818-1
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.