Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bombesin Receptor 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APS302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APS302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APS302 Supplier Invitrogen™ Supplier No. PA3302
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA3-302 detects BRS3 from human samples. PA3-302 has been successfully used in Western blot and immunohistochemistry (paraffin) procedures. By Western blot, this antibody detects a ~50-70 kDa protein representing BRS3. The PA3-302 immunogen is a synthetic peptide corresponding to residues E(381) I S V T S F T G C S V K Q A E D R F(399) of human BRS3 protein.

BRS3 expression has been reported in lung (normal and cancer), nasal mucosa, placenta, and uterus. ESTs have been isolated from normal brain and kidney cancer libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more thana thousandmembers currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Twoblockbuster drugs which have been developed as GPCR-targetedpharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bombesin Receptor 3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene BRS3
Gene Accession No. P32247
Gene Alias BB3; BB3R; bombesin like receptor 3; bombesin receptor subtype 3; bombesin receptor subtype-3; bombesin-like receptor 3; BRS 3; BRS3; BRS-3; G-protein coupled receptor; RP1-196E23.3
Gene Symbols BRS3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues E(381) I S V T S F T G C S V K Q A E D R F(399) of human BRS3 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 680
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.