Learn More
Invitrogen™ Bombesin Receptor 3 Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%); Gibbon (95%); Monkey (90%); Bat, Elephant (85%); Horse, Rabbit, Guinea pig (80%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Specifications
Specifications
| Antigen | Bombesin Receptor 3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | BRS3 |
| Gene Accession No. | P32247 |
| Gene Alias | BB3; BB3R; bombesin like receptor 3; bombesin receptor subtype 3; bombesin receptor subtype-3; bombesin-like receptor 3; BRS 3; BRS3; BRS-3; G-protein coupled receptor; RP1-196E23.3 |
| Gene Symbols | BRS3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.