Learn More
Restek™ Bonded Reversed Phase Resprep SPE Cartridges
SDP
Hydrophobic (nonpolar) silica-based adsorbents; used to extract hydrophobic analytes from polar matrices.
$501.50 - $557.00
Specifications
Adsorbent Type Hydrophobic (non-Polar)
Base Material Silica
Column Phase Reversed Bonded
Sorbent C18 (high load)
Type SPE Cartridge
View More Specs
Products 2
Catalog Number Mfr. No. Tube Size Volume (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Tube Size Volume (Metric) Quantity Price Quantity & Availability  
06-713-095
SDP
View Documents
Restek™
26030
1 mL, 100 mg 1 mL, 100 mg 100/Pk.
Each for $501.50
Only null left
Add to Cart
06-713-097
SDP
View Documents
Restek™
26034
20 mL, 5 g 20 mL, 5 g 20/Pk.
Each for $557.00
Only null left
Add to Cart
Description
Description
  • Manufactured using high density polypropylene
  • May be processed by any of these techniques: positive pressure, sidearm flask, centrifuge or vacuum manifold
Specifications
Specifications
Hydrophobic (non-Polar)
Reversed Bonded
SPE Cartridge
HDPE
Silica
C18 (high load)
Yes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.