Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bone marrow stromal cell antigen 1 (BST1) Monoclonal Antibody (OTI9B2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF812187


Description
Bone marrow stromal cell antigen 1 (BST1) Monoclonal antibody specifically detects Bone marrow stromal cell antigen 1 (BST1) in Human samples. It is validated for Western BlotSpecifications
Bone marrow stromal cell antigen 1 (BST1) | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P25311 | |
Mouse | |
Protein A/G | |
RUO | |
563 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Western Blot | |
OTI9B2 | |
Unconjugated | |
AZGP1 | |
114/A10; A530073F09; ADP-ribosyl cyclase 2; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 2; antigen BP3; Bone marrow stromal antigen 1; bone marrow stromal cell antigen 1; Bp3; Bp-3; BP-3 alloantigen; Bst1; BST-1; Bsta1; cADPr hydrolase 2; CD157; CD157 antigen; Cyclic ADP-ribose hydrolase 2; leukocyte antigen 65; Ly65; Ly-65; NAD(+) nucleosidase; NAD+ nucleosidase | |
Human recombinant protein fragment corresponding to amino acids 21-298 of human AZGP1 (NP_001176) produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction