Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bone marrow stromal cell antigen 1/CD157 Antibody (029), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB229098 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB229098 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB229098 Supplier Novus Biologicals Supplier No. NBP289249AF594
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Bone marrow stromal cell antigen 1/CD157 Monoclonal antibody specifically detects Bone marrow stromal cell antigen 1/CD157 in Human samples. It is validated for ELISA

Specifications

Antigen Bone marrow stromal cell antigen 1/CD157
Applications ELISA
Classification Monoclonal
Clone 029
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias ADP-ribosyl cyclase 2, Bone marrow stromal antigen 1, bone marrow stromal cell antigen 1, BST-1, cADPr hydrolase 2, CD157, CD157 antigen, Cyclic ADP-ribose hydrolase 2, EC 3.2.2.5, NAD(+) nucleosidase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Bone marrow stromal cell antigen 1/CD157 (Met 1-Lys 292; NP_004325.2).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 683
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.