Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bone marrow stromal cell antigen 1/CD157 Antibody (213), DyLight 594 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290531DL594
Description
Bone marrow stromal cell antigen 1/CD157 Monoclonal antibody specifically detects Bone marrow stromal cell antigen 1/CD157 in Mouse samples. It is validated for ELISA, ELISASpecifications
Bone marrow stromal cell antigen 1/CD157 | |
Monoclonal | |
DyLight 594 | |
ADP-ribosyl cyclase 2, Bone marrow stromal antigen 1, bone marrow stromal cell antigen 1, BST-1, cADPr hydrolase 2, CD157, CD157 antigen, Cyclic ADP-ribose hydrolase 2, EC 3.2.2.5, NAD(+) nucleosidase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Bone marrow stromal cell antigen 1/CD157 (NP_033893.2; Met1-Glu285). | |
0.1 mL | |
Immunology | |
683 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
213 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction