Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bone marrow stromal cell antigen 1/CD157 Antibody (534509), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB47361E
Description
Bone marrow stromal cell antigen 1/CD157 Monoclonal antibody specifically detects Bone marrow stromal cell antigen 1/CD157 in Human samples. It is validated for Flow Cytometry, ELISA, CyTOFSpecifications
| Bone marrow stromal cell antigen 1/CD157 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, CyTOF | |
| 534509 | |
| Flow Cytometry, ELISA, CyTOF-ready | |
| ADP-ribosyl cyclase 2, Bone marrow stromal antigen 1, bone marrow stromal cell antigen 1, BST-1, cADPr hydrolase 2, CD157, CD157 antigen, Cyclic ADP-ribose hydrolase 2, EC 3.2.2.5, NAD(+) nucleosidase | |
| NS0 mouse myeloma cell line transfected with human CD157, Gly29-Lys292, Accession # Q10588 | |
| 0.1 mL | |
| Immunology | |
| 683 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction