Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals Bone Morphogenetic Protein 2, Human
Click to view available options
Quantity:
10μg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Formulation | Lyophilized powder |
Quantity | 10μg |
Source | Escherichia coli |
Storage Requirements | -30° to -10°C |
Regulatory Status | RUO |
Product Type | Bone Morphogenetic Protein 2, Human |
Recombinant | Native |
Form | Lyophilized |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction