Learn More
Invitrogen™ Bone SialoProtein Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue.
Specifications
Specifications
| Antigen | Bone SialoProtein |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and 0.05mg sodium azide |
| Gene | Ibsp |
| Gene Accession No. | P13839, P21815, Q61711 |
| Gene Alias | BNSP; Bone sialoprotein 2; bone sialoprotein II; Bsp; BSP II; Bsp2; BSPII; BSP-II; Cell-binding sialoprotein; IBSP; integrin binding sialoprotein; integrin-binding sialoprotein; SP-II |
| Gene Symbols | Ibsp |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.