Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Bonn Scissors, Tungsten
SDP

Catalog No. p-7163519 Shop All Stoelting Co Products
Click to view available options
Model:
Straight, S/S tip
Straight, Strong cut

Tungsten carbide scissors are especially good for cutting cartilage or for many repeated cuts such as in necropsy studies.

  • Tungsten carbide is 5 times harder than stainless steel.
  • Scissors with tungsten carbide inserts along their blades last much longer than stainless steel scissors.

Specifications

For Use With (Application) Neuroscience
Length (English) 3.5 in.
Material Tungsten
Width (Metric) 15 mm blades
Model Straight, S/S tip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.