Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BoNT-B Light Chain Antibody [DyLight 488], Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals AF5420K
Description
BoNT-B Light Chain Polyclonal antibody specifically detects BoNT-B Light Chain in C. botulinum samples. It is validated for Immunoprecipitation,Western BlotSpecifications
BoNT-B Light Chain | |
Polyclonal | |
50mM Sodium Borate | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
6030102 | |
Store at 4°C in the dark. | |
IgG |
Immunoprecipitation, Western Blot | |
DyLight 488 | |
DcD | |
E. coli-derived recombinant Clostridium BoNT-B Light Chain, Pro2-His428, Accession # P10844 | |
0.1 mL | |
Primary | |
C. botulinum | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction