Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BoNT-E Heavy Chain Sheep anti-C. botulinum, Polyclonal, R&D Systems™

Catalog No. AF7135SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7135SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7135SP Supplier R&D Systems Supplier No. AF7135SP

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Sheep Polyclonal Antibody

BoNT-E Heavy Chain Polyclonal antibody AF7135SP specifically detects BoNT-E Heavy Chain in C. botulinum samples. It is validated for Western Blotting

Specifications

Antigen BoNT-E Heavy Chain
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P30995
Host Species Sheep
Immunogen E. coli-derived recombinant C. botulinum BoNT-E Heavy Chain, Lys845-Lys1251, Accession # P30995
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects C. botulinum BoNT-E Heavy Chain in direct ELISAs and Western blots. In direct ELISAs, approximately 4% cross-reactivity with BoNT-G/HCR and BoNT-A/HCR is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species C. botulinum
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.