Learn More
Geoblox Books

Description
Designed by a teacher, Geoblox books allow each student to create and study their own models. Once you purchase a book, you can copy as many model patterns as needed, time and time again.
Tectonics Block Models:
Contains 17 block model patterns. The first 9 can be connected to form a 45 in. long series of plate boundaries.
Note:
When you purchase this product you will receive a book of patterns you will make copies from. Give your students a copy of each pattern and let them make their own models.
Specifications
Specifications
Format | Book |
Product Type | Tectonics Block Models |
Includes | 17 Block model patterns. The first 9 can be connected to form a 45 in. long series of plate boundaries |
Subject | Social Science |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.