Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Geoblox Books

Click to view available options
Product Type:
Oceanography Block Models
Description
Designed by a teacher, Geoblox books allow each student to create and study their own models. Once you purchase a book, you can copy as many model patterns as needed, time and time again.
Oceanography Block Models:
Contains 20 patterns ranging from Barrier Islands to the marine carbon cycle.
Note:
When you purchase this product you will receive a book of patterns you will make copies from. Give your students a copy of each pattern and let them make their own models.
Specifications
Specifications
| Format | Book |
| Product Type | Oceanography Block Models |
| Includes | 20 Patterns ranging from barrier islands to the marine Carbon cycle |
| Subject | Social Science |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction