Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BOP
GREENER_CHOICESDP

Catalog No. 604710 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-471-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-471-0 Supplier Tocris Bioscience™ Supplier No. 6047/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dual α9β1/α4β1 integrin inhibitor; preferentially mobilizes HSCs

BOP is a dual α9β1/α4β1 integrin inhibitor; rapidly and preferentially mobilizes HSCs and progenitors. Augments HSC mobilization in combination with AMD3100 (Cat. No. 3299).

Fluorogenic, bright and photostable Fluorescent Conjugate BOP-JF646 (Cat. No. 6997) also available.

High affinity fluorescent derivative R-BC154 (Cat. No. 6048) also available.

Specifications

CAS 1947348-42-6
Quantity 10 mg
Solubility Soluble to 100mM in water and to 100mM in DMSO
Molecular Formula C25H28N3O7SNa
Formula Weight 537.56
Color White
Product Type Inhibitor

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.