Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BOP1 Antibody (T7-P1B7*A8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250124H
Description
BOP1 Monoclonal specifically detects BOP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BOP1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
BOP1 | |
Peptide Sequence - KGHVLTRDLG | |
0.1 mL | |
Epigenetics | |
23246 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
T7-P1B7*A8 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
block of proliferation 1, KIAA0124Block of proliferation 1 protein, ribosome biogenesis protein BOP1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction