Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BOP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117811
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117811 Supplier Invitrogen™ Supplier No. PA5117811
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BOP1 Polyclonal Antibody for Western Blot, ICC/IF, IP

Component of the PeBoW complex, which is required for maturation of 28S and 5.8S ribosomal RNAs and formation of the 60S ribosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BOP1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene BOP1
Gene Accession No. Q14137
Gene Alias AU020183; AW146150; block of proliferation 1; block of proliferation 1 protein; block of proliferation 1 protein {ECO:0000255; Bop1; D18861; Erb1p; HAMAP-Rule:MF_03027}; Kiaa0124; mKIAA0124; ribosome biogenesis protein BOP1; ribosome biogenesis protein BOP1 {ECO:0000255
Gene Symbols BOP1
Host Species Rabbit
Immunogen E. coli-derived Human BOP1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23246
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.