Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Bora Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB68561SP
This item is not returnable.
View return policy
Description
Bora Monoclonal specifically detects Bora in Human samples. It is validated for Western Blot.Specifications
Bora | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
Q6PGQ7 | |
BORA | |
E. coli-derived recombinant human Bora Gly2-Ala180 Accession # Q6PGQ7 | |
25 μg | |
Primary | |
Detects human Bora in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
694746 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
aurora borealis, BORA, C13orf34, chromosome 13 open reading frame 34, FLJ22624, hsBora, protein aurora borealis | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
79866 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction