Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bora Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589480
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589480 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589480 Supplier Invitrogen™ Supplier No. PA589480
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KYSLGSITSP SPISSPTFSP IEFQIGETPL SEQRKFTVHS PDASSGTNSN GITNPCIRSP YIDGCSPIKN WSPMRLQMYS GGTQYRTSVI QIPFTLETQG EDEEDKENIP STDVSSPAMD AAGIHLRQFS NEASTHGTHL VVTAMSVTQN QSSASEKELA LLQDVEREKD NNTVDMVDPI EIADETTWIK EPVDNGSLPM TDFVSGIAFS IENSHMCMSP LAESSVIPCE SSNIQMDSGY NTQNCGSNIM DTVGAESYCK ESDAQTCEVE SKSQAFNMKQ DHTTQRCWMK TASPFQCSSP; Positive Samples: HT-29, HeLa.

Bora (Protein aurora borealis) is a key activator of Aurora Related Protein Kinase A (ARK-1), which is a centrosome-associated serine/threonine kinase that regulates centrosome maturation, bipolar spindle assembly and chromosome segregation during mitosis. Bora is localized to the nucleus until mitosis is initiated, then translocates to the cytoplasm in a Cdc2 dependent manner. Activation of Cdc2 initiates the release of Bora into the cytoplasm where it can bind and activate ARK-1. PLK1 (polo-like kinase-1) interacts with Bora to control the accessibility of its activation loop for phosphorylation and activation by ARK-1. Bora and ARK-1 cooperatively activate PLK1 and control mitotic entry. Bora mutants result in multipolar spindles in mitosis identical to those observed when ARK-1 function is blocked. Thus, the ARK1-Bora-PLK1 regulatory circuit in mammalian cells elucidates a key mechanism in cell cycle regulation. At least three isoforms of Bora are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bora
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene BORA
Gene Accession No. Q6PGQ7
Gene Alias 6720463M24Rik; AI317232; aurora borealis; BORA; bora, aurora kinase A activator; C13orf34; HsBora; Protein aurora borealis; RGD1309522; RP11-342J4.2
Gene Symbols BORA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 260-559 of human BORA (NP_079084.4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79866
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.