Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Bora Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114916
Description
Antibody detects endogenous levels of total BORA.
Bora (Protein aurora borealis) is a key activator of Aurora Related Protein Kinase A (ARK-1), which is a centrosome-associated serine/threonine kinase that regulates centrosome maturation, bipolar spindle assembly and chromosome segregation during mitosis. Bora is localized to the nucleus until mitosis is initiated, then translocates to the cytoplasm in a Cdc2 dependent manner. Activation of Cdc2 initiates the release of Bora into the cytoplasm where it can bind and activate ARK-1. PLK1 (polo-like kinase-1) interacts with Bora to control the accessibility of its activation loop for phosphorylation and activation by ARK-1. Bora and ARK-1 cooperatively activate PLK1 and control mitotic entry. Bora mutants result in multipolar spindles in mitosis identical to those observed when ARK-1 function is blocked. Thus, the ARK1-Bora-PLK1 regulatory circuit in mammalian cells elucidates a key mechanism in cell cycle regulation. At least three isoforms of Bora are known to exist.
Specifications
| Bora | |
| Polyclonal | |
| Unconjugated | |
| BORA | |
| 6720463M24Rik; AI317232; aurora borealis; BORA; bora, aurora kinase A activator; C13orf34; HsBora; Protein aurora borealis; RGD1309522; RP11-342J4.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 77744, 79866 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q6PGQ7, Q8BS90 | |
| BORA | |
| A synthesized peptide derived from human BORA(Accession Q6PGQ7), corresponding to amino acid residues H474-Q524. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction