Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Borane-dimethyl sulfide complex, 2M in THF, packaged under Argon in resealable ChemSeal™ bottles
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 0.855 g/mL |
| Flash Point | −17°C (−1°F) |
| Odor | Characteristic |
| Packaging | ChemSeal™ bottles |
| Linear Formula | (CH3)2S·BH3 |
| Quantity | 100 mL |
| UN Number | UN3399 |
| Beilstein | 3663489 |
| Sensitivity | Air and moisture sensitive |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H225-H260-H302-H315-H318-H335-H336-H351 |
| missing translation for 'healthHazard2' | GHS H Statement H225-H261-H318-H302-H335-H315 Highly flammable liquid and vapor. In contact with water releases flammable gas. Causes serious eye damage. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. |
| missing translation for 'healthHazard3' | P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P335+P334 |
| DOTInformation | Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction