Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Borane-dimethylamine complex, 97%

Manufacturer:  Alfa Aesar™ 8914914

Quantity 25g

 View more versions of this product

Catalog No. AA8914914


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

74-94-2
58.92
UN2925
Borane-dimethylamine complex
BH3·NH(CH3)2
MFCD00051068
Dimethylamine borane

Specifications

74-94-2
Crystalline
58.92
UN2925
White
59° to 65°C (1° to 2mmHg)
Air & Moisture Sensitive
97%
0.69g/mL
BH3·NH(CH3)2
MFCD00051068
36°C
Dimethylamine borane
25g
Borane-dimethylamine complex

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H310-H330-H314-H318-H228
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.

RTECSNumber : IP9450000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit