Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-dimethylamine complex, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
74-94-2
C2H10BN
58.92
Dimethylamineborane, DMAB
74-94-2
C2H10BN
58.92
Dimethylamineborane, DMAB
Reduction reagent
Flash Point 65°C
Melting Point 33.0°C to 37.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Color White to Yellow
Assay Percent Range 98+%
Linear Formula (CH3)2NH·BH3
Solubility Information Solubility in water: 125g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,thf,,diglyme,benzene,toluene,chlorobenzene
Formula Weight 58.92
Physical Form Flakes or Granules
Percent Purity 98+%
Chemical Name or Material Borane-dimethylamine complex
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 200-823-7