Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane dimethyl sulfide complex, 2M solution in THF, AcroSeal™

Catalog No. AC185078000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 13292-87-0 | C2H9BS | 75.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Borane-methyl sulfide complex
Color Colorless to Yellow
Name Note 2M Solution in Tetrahydrofuran
CAS 13292-87-0, 109-99-9, 75-18-3
Molecular Formula C2H9BS
Linear Formula (CH3)2S·BH3
MDL Number MFCD00013189
Concentration 1.80 to 2.20M
Fieser 04,191; 05,47; 06,64; 08,49; 10,49; 14,52; 17,50
Density 0.8500g/mL
Merck Index 15, 1336
Synonym dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ethyl ether, tetrahydrofurane, hexane,, toluene, methylene chloride and other aprotic, solvents
InChI Key RMHDLBZYPISZOI-UHFFFAOYSA-N
SMILES B.CSC
IUPAC Name (methylsulfanyl)methane borane
Molecular Weight (g/mol) 75.96
PubChem CID 9833925
Formula Weight 75.95
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.85
Quantity 800 mL
Flash Point −17°C
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.