Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™

Catalog No. p-4813360
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle

CAS: 13292-87-0 | C2H9BS | 75.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Borane dimethyl sulfide complex
CAS 13292-87-0, 108-88-3
Quantity 100 mL
Molecular Formula C2H9BS
Linear Formula (CH3)2S·BH3
MDL Number MFCD00013189
Flash Point 7°C
Merck Index 15,1336
Synonym dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
InChI Key RMHDLBZYPISZOI-UHFFFAOYSA-N
SMILES B.CSC
IUPAC Name boron;methylsulfanylmethane
Molecular Weight (g/mol) 75.96
PubChem CID 9833925
Formula Weight 75.95
Concentration or Composition (by Analyte or Components) 1.90 to 2.20 M
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.856
Density 0.8560g/mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.