Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Borane-pyridine complex, 95%
Click to view available options
Quantity:
25 mL
100 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 10.0°C to 11.0°C |
| Density | 0.9200g/mL |
| Flash Point | 73°C |
| Assay Percent Range | 95% |
| Packaging | Glass bottle |
| Refractive Index | 1.5300 to 1.5320 |
| Quantity | 25 mL |
| Fieser | 08,50; 09,59; 12,65 |
| Specific Gravity | 0.92 |
| Solubility Information | Solubility in water: reacts slowly with water. Other solubilities: soluble in methanol,ethanol |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal in contact with skin. Fatal if inhaled. Toxic if swallowed. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Gently wash with plenty of soap and water. |
| EINECSNumber | 203-773-4 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction