Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borate, 0.5 M Buffer Solution, pH 8.0, PROMO, Thermo Scientific Chemicals

Catalog No. J63742APPM
Click to view available options
Quantity:
500 mL

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21 g/mol
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Concentration 0.5 M
pH 8.0
Physical Form Liquid
Quantity 500 mL
Chemical Name or Material Borate buffer solution

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.