Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bordetella pertussis LOS-A Monoclonal Antibody (D26E)
GREENER_CHOICE

Catalog No. MA17005
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17005 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17005 Supplier Invitrogen™ Supplier No. MA17005
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7005 detects Bordetella pertussis. This antibody is specific for LOS-A of Bordetella pertussis and B. bronchiseptica. MA1-7005 has been successfully used in ELISA, immunofluorescence and Western blot procedures. The MA1-7005 immunogen is whole B. pertussis lysate.

Bordetella lipopolysaccharides (LPS) are characterized by the absence of the O-polysaccharide (O-antigen) but have an extended core region and are more commonly termed as LOS. B. pertussis is a non-motile, gram-negative bacteria that is the common causative agent for whooping cough. B. bronchiseptica is a closely related species that is often involved in respiratory tract infection in smaller mammals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bordetella pertussis LOS-A
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone D26E
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias B. pertussis; LOS-A
Host Species Mouse
Immunogen Whole Bordetella pertussis lysate.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.