Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bordetella Pertussis Toxin Antibody (1280/204), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064514AF488
Description
Bordetella Pertussis Toxin Monoclonal specifically detects Bordetella Pertussis Toxin in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Bordetella Pertussis Toxin | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
NB100-64514 is reactive with Bordetella pertussis toxin in a simple ELISA. Reactivities with other organisms have not yet been determined. | |
Bacteria | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence | |
1280/204 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
pertussis toxin, pertussis toxin A protomer | |
Native | |
0.1 mL | |
Primary | |
Protein A purified | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction