Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bordetella Pertussis Toxin Antibody (1280/204), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064514AF750
Description
Bordetella Pertussis Toxin Monoclonal specifically detects Bordetella Pertussis Toxin in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Bordetella Pertussis Toxin | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
NB100-64514 is reactive with Bordetella pertussis toxin in a simple ELISA. Reactivities with other organisms have not yet been determined. | |
Store at 4°C in the dark. |
ELISA, Immunocytochemistry, Immunofluorescence | |
1280/204 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
pertussis toxin, pertussis toxin A protomer | |
Native | |
Primary | |
Bacteria | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction