Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bordetella pertussis Toxin Monoclonal Antibody (1280/204)
GREENER_CHOICE

Catalog No. PIMA183016
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183016 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183016 Supplier Invitrogen™ Supplier No. MA183016
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Bordetella Pertussis Toxin antibody, clone 1280/204 is reactive with Bordetella pertussis toxin in a simple ELISA. Reactivities with other organisms have not yet been determined.

B. pertussis has the ability to inhibit the function of the host's immune system. The toxin, known as pertussis toxin (or PTx), inhibits G protein coupling that regulates an adenylate cyclase-mediated conversion of ATP to cyclic AMP. The end result is phagocytes convert too much ATP to cyclic AMP, which can cause disturbances in cellular signaling mechanisms, and prevent phagocytes from correctly responding to an infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bordetella pertussis Toxin
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1280/204
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias B. pertussis; PTx
Host Species Mouse
Immunogen Native protein.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.