Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Borealin Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP122832APC
Description
Borealin Polyclonal antibody specifically detects Borealin in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
Borealin | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
BOR, BOREALIN, cell division cycle associated 8, Cell division cycle-associated protein 8, Dasra B, dasra-B, DasraB, FLJ10468, FLJ12042, hDasra-B, MESRGP, PESCRG3, Pluripotent embryonic stem cell-related gene 3 protein | |
Synthetic peptide made to an internal portion of human Borealin (within residues 200-250). [Swiss-Prot# Q53HL2] | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers, Zinc Finger | |
55143 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction