Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 19 publications
$206.50 - $499.50
Specifications
| Antigen | BMP-2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 ug/ml, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 30700748), Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP119751SS
![]() |
Novus Biologicals
NBP119751SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP119751
![]() |
Novus Biologicals
NBP119751 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
BMP-2 Polyclonal specifically detects BMP-2 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
| BMP-2 | |
| Western Blot 0.5 ug/ml, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 30700748), Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Embryonic Stem Cell Markers, Stem Cell Signaling Pathway | |
| PBS (pH 7.4) with 0.05% Sodium Azide | |
| BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2 | |
| BMP2 | |
| IgG | |
| Affinity Purified | |
| 44 kDa |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Canine | |
| P12643, P21274 | |
| 650 | |
| A synthetic peptide made to an internal region of human BMP2 (within residues 250-350) [Swiss-Prot# P12643] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title