Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Borealin Antibody (OTI4G3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271988
Description
Borealin Monoclonal specifically detects Borealin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Borealin | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
BOR, BOREALIN, cell division cycle associated 8, Cell division cycle-associated protein 8, Dasra B, DasraB, Dasra-B, FLJ10468, FLJ12042, hDasra-B, MESRGP, PESCRG3, Pluripotent embryonic stem cell-related gene 3 protein | |
Mouse | |
Affinity Purified | |
Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers, Zinc Finger | |
55143 | |
Human | |
Lyophilized |
Western Blot, Immunohistochemistry | |
OTI4G3 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
CDCA8 | |
Full length human recombinant protein of human CDCA8 (NP_060571) produced in E.coli. | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction