Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borealin Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE1961MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1961MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1961MI Supplier MilliporeSigma Supplier No. ABE1961
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Borealin Antibody is a highly specific rabbit polyclonal antibody that targets Borealin and has been tested in Immunofluorescence, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Borealin
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CDCA8,PESCRG3
Host Species Rabbit
Immunogen A His-tagged full length human recombinant Borealin.
Purification Method Unpurified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_060571
Test Specificity This polyclonal antibody detects Borealin in nocodozole treated HeLa cells.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.