Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, 2% (w/v) with Indicator (Methylene Blue-Methyl Red) for Nitrogen (Ammonia), Nitrogen (Organic), Certified, LabChem™

Catalog No. LC117701
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC117701 500 mL
LC117702 1 L
LC117704 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC117701 Supplier LabChem, Inc. Supplier No. LC117701
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10043-35-3 , 7732-18-5 , 64-17-5 , 64-17-5 , 67-63-0
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

CAS Max % 2, 96, 1, 0.9, 0.052
Linear Formula H3BO3
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 61.83
Grade Certified
Packaging Poly Bottle
Suitability Passes Test
Decomposition Information Boron; Carbon monoxide; Carbon dioxide
Color Purple
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Boric Acid, 2% (w/v) with Indicator (Methylene Blue-Methyl Red) for Nitrogen (Ammonia), Nitrogen (Organic)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May damage fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.