Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boric Acid, 4% (w/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Specifications
Specifications
| Appearance of Solution | Colorless Liquid |
| CAS Min % | 94.0, 3.77 |
| CAS Max % | 98.0, 3.93 |
| Packaging | Natural Poly Bottle |
| Quantity | 1 L |
| Synonym | orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe |
| Percent Purity | 4% W/V |
| Grade | Laboratory |
| Chemical Name or Material | Boric Acid Solution, 4%(w/v) Aqueous Solution |
Safety and Handling
| ShelfLife | 24 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction