Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCB050025A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCB050025A 2.5 kg
RDCB0500500 500 g
RDCB0500100 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RDCB050025A Supplier Ricca Chemical Company Supplier No. RDCB05002.5A5
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
IUPAC Name boric acid
SMILES OB(O)O

Specifications

CAS Max % 100
Heavy Metals (as Pb) max. 0.001%
Non_volatile Matter max. 0.05%
Packaging Natural Poly Bottle
Quantity 2.5 kg
Percent Purity min 99.5%
Grade ACS
Chemical Name or Material Boric Acid
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.