Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boric Acid Enhanced Specimen Container with 5 Tablets, Cenmed™

Click to view available options
Quantity:
100 Containers, 5 x 2 g Tablets
Description
- Inclusion of five 2 g tablets of boric acid provides an advanced solution, elevating the container's capabilities for preserving various types of specimens
- Tailored for non-sterile environments, this container is designed to meet the specific needs of various applications, providing a reliable option for sample storage
- Stand out in your preservation practices with this enhanced specimen container, delivering ideal results with the added benefits of boric acid
Specifications
Specifications
Physical Form | Tablets, Liquid |
CAS | 10043-35-3 |
Color | Clear |
Quantity | 100 Containers, 5 x 2 g Tablets |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction