Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5391413 Shop All Avantor Products
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Packaging Poly Bottle
Specific Gravity 1L = 1.43kg
Quantity 500 g
Physical Form Granular
Formula Weight 61.83
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Boric Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.