Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Granular, Technical, Spectrum™ Chemical
SDP

Catalog No. 18609172 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-172 100 lb. Fiber Drum
18-609-175 500 g Fiber Drum
18-609-174 2.5 kg Fiber Drum
18-609-173 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-609-172 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1133100LBBL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10043-35-3
Molecular Weight (g/mol) 61.83

Specifications

CAS Max % 1
Packaging Fiber Drum
Quantity 100 lb.
Grade Technical
Chemical Name or Material Boric Acid

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.