Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Boric acid, Molecular Biology Grade, 99.5% min

Click to view available options
Quantity:
500g
1kg
5kg
10043-35-3
BH3O3
61.831
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe
7628
CHEBI:33118
boric acid
B(O)(O)O
10043-35-3
BH3O3
61.831
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe
7628
CHEBI:33118
boric acid
B(O)(O)O
Quantity 500g
Color White
pH 3.8 to 4.8
Beilstein 1697939
Merck Index 14,1336
Density 1.435g/mL
Formula Weight 61.83
Physical Form Powder
Percent Purity ≥99.5%
Grade Molecular Biology
Odor Odorless
Melting Point ~185°C (decomposition)
Chemical Name or Material Boric acid
Show More Show Less

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

Danger

RTECSNumber : ED4550000

TSCA : TSCA